siRNA Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769281.1 | XP_008767503.1 | 22, 23 | 6254 | ||
| XM_008769282.1 | XP_008767504.1 | 22, 23 | 6248 | |||
| XM_008769283.1 | XP_008767505.1 | 21, 22 | 6199 | |||
| XM_008769284.1 | XP_008767506.1 | 21, 22 | 6193 | |||
| XM_008769285.1 | XP_008767507.1 | 22, 23 | 6253 | |||
| XM_008769286.1 | XP_008767508.1 | 22, 23 | 6247 | |||
| XM_008769287.1 | XP_008767509.1 | 22, 23 | 6254 | |||
| XM_008769288.1 | XP_008767510.1 | 21, 22 | 6199 | |||
| XM_008769292.1 | XP_008767514.1 | 21, 22 | 6044 | |||
| XM_008769294.1 | XP_008767516.1 | 21, 22 | 6026 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009125, AK135879, AK135976, AF041472.

Atxn2
Genomic Map