siRNA Details
| Gene Symbol: Anapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080147.1 | NP_001073616.1 | 13 | 1607 | ||
| XM_006249363.1 | XP_006249425.1 | 13 | 1668 | |||
| GenBank mRNA | BC129106.1 | - | 13 | 1607 | 7100647 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anapc5| Gene Symbol: Anapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080147.1 | NP_001073616.1 | 13 | 1607 | ||
| XM_006249363.1 | XP_006249425.1 | 13 | 1668 | |||
| GenBank mRNA | BC129106.1 | - | 13 | 1607 | 7100647 | |