siRNA Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011904.1 | NP_001011904.1 | 4 | 1065 | ||
| XM_006249434.2 | XP_006249496.1 | 4 | 642 | |||
| GenBank mRNA | BC083831.1 | - | 4 | 1065 | 7112480 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf10| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011904.1 | NP_001011904.1 | 4 | 1065 | ||
| XM_006249434.2 | XP_006249496.1 | 4 | 642 | |||
| GenBank mRNA | BC083831.1 | - | 4 | 1065 | 7112480 | |