siRNA Details
| Gene Symbol: Rbms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025403.1 | NP_001020574.1 | 9 | 938 | ||
| XM_006240742.2 | XP_006240804.1 | 13 | 1171 | |||
| XM_006240744.2 | XP_006240806.1 | 11 | 1340 | |||
| XM_008765021.1 | XP_008763243.1 | 12 | 1427 | |||
| XM_008765022.1 | XP_008763244.1 | 11 | 1347 | |||
| XM_008765023.1 | XP_008763245.1 | 10 | 1164 | |||
| GenBank mRNA | BC097377.1 | - | 9 | 938 | 7445017 | |

Rbms2
Genomic Map