siRNA Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004206.1 | NP_001004206.1 | 2 | 275 | ||
| GenBank mRNA | BC079095.1 | - | 2 | 275 | 7124036 | |
| FQ233061.1 | - | 2 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pa2g4| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004206.1 | NP_001004206.1 | 2 | 275 | ||
| GenBank mRNA | BC079095.1 | - | 2 | 275 | 7124036 | |
| FQ233061.1 | - | 2 | 283 | |||