siRNA Details
| Gene Symbol: Neurod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105942.1 | NP_001099412.1 | 2 | 542 | ||
| XM_006240818.2 | XP_006240880.1 | 4 | 932 | |||
| XM_006240819.2 | XP_006240881.1 | 4 | 1065 | |||
| XM_006240820.2 | XP_006240882.1 | 3 | 844 | |||
| XM_006240822.2 | XP_006240884.1 | 4 | 808 | |||
| XM_006240823.2 | XP_006240885.1 | 2 | 578 | |||

Neurod4
Genomic Map