siRNA Details
| Gene Symbol: Olr1059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001362.1 | NP_001001362.1 | 1 | 491 | ||
| Gene Symbol: LOC103690258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765077.1 | XP_008763299.1 | 1 | 491 | ||

Olr1059
Genomic Map