siRNA Details
Gene Symbol: Olr1059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001362.1 | NP_001001362.1 | 1 | 269 |
Gene Symbol: LOC103690258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765077.1 | XP_008763299.1 | 1 | 269 |
