siRNA Details
| Gene Symbol: Olr1058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000066.1 | NP_001000066.1 | 1 | 731 | ||
| Gene Symbol: LOC103692758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765057.1 | XP_008763279.1 | 1 | 1078 | ||

Olr1058
Genomic Map