siRNA Details
Gene Symbol: Asna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100505.1 | NP_001093975.1 | 6 | 870 | ||
XM_008772367.1 | XP_008770589.1 | 7 | 1691 | |||
XM_008772368.1 | XP_008770590.1 | 7 | 1819 | |||
XM_008772369.1 | XP_008770591.1 | 7 | 1814 | |||
GenBank mRNA | BC098819.1 | - | 6 | 805 |
