siRNA Details
Gene Symbol: Asna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100505.1 | NP_001093975.1 | 4 | 490 | ||
XM_008772367.1 | XP_008770589.1 | 5 | 1311 | |||
XM_008772368.1 | XP_008770590.1 | 5 | 1439 | |||
XM_008772369.1 | XP_008770591.1 | 5 | 1434 | |||
GenBank mRNA | BC098819.1 | - | 4 | 425 |
