siRNA Details
| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081634.1 | NP_001075103.1 | 6 | 1061 | ||
| GenBank mRNA | BC166590.1 | - | 5 | 1078 | 7938435 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw9| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081634.1 | NP_001075103.1 | 6 | 1061 | ||
| GenBank mRNA | BC166590.1 | - | 5 | 1078 | 7938435 | |