siRNA Details
| Gene Symbol: Insig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178091.4 | NP_835192.1 | 3 | 447 | ||
| XM_006249667.2 | XP_006249729.1 | 4 | 912 | |||
| XM_006249668.2 | XP_006249730.1 | 4 | 760 | |||
| XM_006249669.2 | XP_006249731.1 | 4 | 727 | |||
| GenBank mRNA | AY152392.1 | - | 3 | 447 | ||
| BC085682.1 | - | 4 | 585 | 7189822 | ||

Insig2
Genomic Map