siRNA Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191669.1 | NP_001178598.1 | 14 | 2999 | ||
| XM_008769435.1 | XP_008767657.1 | 15 | 7387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thsd7b| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191669.1 | NP_001178598.1 | 14 | 2999 | ||
| XM_008769435.1 | XP_008767657.1 | 15 | 7387 | |||