siRNA Details
| Gene Symbol: Pik3c2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105951.1 | NP_001099421.1 | 2 | 571 | ||
| XM_006249754.2 | XP_006249816.1 | 2 | 638 | |||
| XM_008769442.1 | XP_008767664.1 | 2 | 1478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3c2b| Gene Symbol: Pik3c2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105951.1 | NP_001099421.1 | 2 | 571 | ||
| XM_006249754.2 | XP_006249816.1 | 2 | 638 | |||
| XM_008769442.1 | XP_008767664.1 | 2 | 1478 | |||