siRNA Details
| Gene Symbol: Tmem183a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013871.1 | NP_001013893.1 | 7 | 942 | ||
| XM_006249849.2 | XP_006249911.1 | 7 | 1006 | |||
| XM_006249850.2 | XP_006249912.1 | 6 | 930 | |||
| GenBank mRNA | BC079375.1 | - | 7 | 942 | 7110239 | |
| FQ210515.1 | - | 7 | 975 | |||

Tmem183a
Genomic Map