siRNA Details
| Gene Symbol: Tmem183a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013871.1 | NP_001013893.1 | 6 | 765 | ||
| XM_006249849.2 | XP_006249911.1 | 6 | 829 | |||
| GenBank mRNA | BC079375.1 | - | 6 | 765 | 7110239 | |
| FQ210515.1 | - | 6 | 798 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_020588, NM_001042485, AB041592, AK004922, AK152852, and others.

Tmem183a
Genomic Map