siRNA Details
| Gene Symbol: Adipor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207587.1 | NP_997470.1 | 7 | 1043 | ||
| XM_006249852.2 | XP_006249914.1 | 7 | 1079 | |||
| XM_008769543.1 | XP_008767765.1 | 7 | 1380 | |||
| XM_008769544.1 | XP_008767766.1 | 7 | 1742 | |||
| GenBank mRNA | BC061838.1 | - | 7 | 1043 | 5599938 | |
| DQ148391.1 | - | 6 | 831 | |||

Adipor1
Genomic Map