siRNA Details
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105953.2 | NP_001099423.2 | 4, 5 | 339 | ||
NM_001277263.1 | NP_001264192.1 | 4, 5 | 416 | |||
GenBank mRNA | BC169086.1 | - | 3, 4 | 306 | 5621176 |
