siRNA Details
| Gene Symbol: Tmem9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105953.2 | NP_001099423.2 | 5 | 430 | ||
| NM_001277263.1 | NP_001264192.1 | 5 | 507 | |||
| GenBank mRNA | BC169086.1 | - | 4 | 397 | 5621176 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem9| Gene Symbol: Tmem9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105953.2 | NP_001099423.2 | 5 | 430 | ||
| NM_001277263.1 | NP_001264192.1 | 5 | 507 | |||
| GenBank mRNA | BC169086.1 | - | 4 | 397 | 5621176 | |