siRNA Details
| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105955.2 | NP_001099425.2 | 18 | 6621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aspm| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105955.2 | NP_001099425.2 | 18 | 6621 | ||