siRNA Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047084.1 | NP_001040549.1 | 5 | 621 | ||
| XM_008769548.1 | XP_008767770.1 | 5 | 647 | |||
| GenBank mRNA | AY651776.1 | - | 5 | 621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs18| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047084.1 | NP_001040549.1 | 5 | 621 | ||
| XM_008769548.1 | XP_008767770.1 | 5 | 647 | |||
| GenBank mRNA | AY651776.1 | - | 5 | 621 | ||