siRNA Details
| Gene Symbol: Edem3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191671.1 | NP_001178600.1 | 16 | 1879 | ||
| XM_006249985.2 | XP_006250047.1 | 16 | 2316 | |||
| XM_006249986.2 | XP_006250048.1 | 16 | 2316 | |||
| XM_006249987.2 | XP_006250049.1 | 16 | 2313 | |||
| XM_008769612.1 | XP_008767834.1 | 16 | 2316 | |||

Edem3
Genomic Map