siRNA Details
Gene Symbol: Shcbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105961.2 | NP_001099431.1 | 5 | 979 | ||
GenBank mRNA | BC160910.1 | - | 5 | 979 | 7456157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Shcbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105961.2 | NP_001099431.1 | 5 | 979 | ||
GenBank mRNA | BC160910.1 | - | 5 | 979 | 7456157 |