siRNA Details
| Gene Symbol: Tdrd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134739.1 | NP_001128211.1 | 2 | 436 | ||
| NM_001134740.1 | NP_001128212.1 | 2 | 436 | |||
| XM_006250037.2 | XP_006250099.1 | 3 | 700 | |||
| XM_006250038.2 | XP_006250100.1 | 3 | 518 | |||
| XM_008769615.1 | XP_008767837.1 | 3 | 487 | |||
| XM_008769616.1 | XP_008767838.1 | 3 | 654 | |||
| GenBank mRNA | BC168218.1 | - | 2 | 436 | 7319314 | |

Tdrd5
Genomic Map