siRNA Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134501.1 | NP_001127973.1 | 7 | 1993 | ||
| XM_006250152.2 | XP_006250214.1 | 8 | 1921 | |||
| XM_008769622.1 | XP_008767844.1 | 7 | 2023 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl13| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134501.1 | NP_001127973.1 | 7 | 1993 | ||
| XM_006250152.2 | XP_006250214.1 | 8 | 1921 | |||
| XM_008769622.1 | XP_008767844.1 | 7 | 2023 | |||