siRNA Details
| Gene Symbol: Kifap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105964.1 | NP_001099434.1 | 8, 9 | 1233 | ||
| XM_008769623.1 | XP_008767845.1 | 9, 10 | 1365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kifap3| Gene Symbol: Kifap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105964.1 | NP_001099434.1 | 8, 9 | 1233 | ||
| XM_008769623.1 | XP_008767845.1 | 9, 10 | 1365 | |||