siRNA Details
| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077643.1 | NP_001071111.1 | 1 | 84 | ||
| XM_006250126.2 | XP_006250188.1 | 1 | 879 | |||
| XM_006250127.2 | XP_006250189.1 | 1 | 879 | |||
| GenBank mRNA | FQ215073.1 | - | 1 | 225 | ||
| FQ216993.1 | - | 1 | 122 | |||
| FQ217960.1 | - | 1 | 153 | |||
| FQ222522.1 | - | 1 | 220 | |||
| FQ224252.1 | - | 1 | 116 | |||

Mpc2
Genomic Map