siRNA Details
| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105967.2 | NP_001099437.2 | 2 | 338 | ||
| XM_008769728.1 | XP_008767950.1 | 2 | 231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmx1a| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105967.2 | NP_001099437.2 | 2 | 338 | ||
| XM_008769728.1 | XP_008767950.1 | 2 | 231 | |||