siRNA Details
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011907.1 | NP_001011907.1 | 11 | 1278 | ||
XM_003751277.2 | XP_003751325.1 | 11 | 1291 | |||
GenBank mRNA | BC082067.1 | - | 11 | 1278 | 7132777 | |
FQ215922.1 | - | 11 | 1282 |
