siRNA Details
| Gene Symbol: Ncstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174864.3 | NP_777353.1 | 15 | 1860 | ||
| XM_006250283.1 | XP_006250345.1 | 14 | 1715 | |||
| XM_008769733.1 | XP_008767955.1 | 15 | 1845 | |||
| XM_008769734.1 | XP_008767956.1 | 15 | 1802 | |||
| XM_008769735.1 | XP_008767957.1 | 13 | 1563 | |||
| GenBank mRNA | AF510722.1 | - | 14 | 1644 | ||
| AY101378.1 | - | 15 | 1768 | |||
| BC074006.1 | - | 16 | 1975 | |||
| BC167747.1 | - | 14 | 1715 | 8365089 | ||
| FQ212793.1 | - | 3 | 263 | |||

Ncstn
Genomic Map