siRNA Details
| Gene Symbol: Olr1584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000081.1 | NP_001000081.1 | 1 | 491 | ||
| XM_006250315.2 | XP_006250377.1 | 3 | 1082 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1584| Gene Symbol: Olr1584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000081.1 | NP_001000081.1 | 1 | 491 | ||
| XM_006250315.2 | XP_006250377.1 | 3 | 1082 | |||