siRNA Details
| Gene Symbol: Olr1588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000082.1 | NP_001000082.1 | 1 | 571 | ||
| XM_008769782.1 | XP_008768004.1 | 1 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1588| Gene Symbol: Olr1588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000082.1 | NP_001000082.1 | 1 | 571 | ||
| XM_008769782.1 | XP_008768004.1 | 1 | 584 | |||