siRNA Details
| Gene Symbol: Spta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011908.3 | NP_001011908.2 | 30 | 4438 | ||
| GenBank mRNA | AY238342.1 | - | 30 | 4438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spta1| Gene Symbol: Spta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011908.3 | NP_001011908.2 | 30 | 4438 | ||
| GenBank mRNA | AY238342.1 | - | 30 | 4438 | ||