siRNA Details
| Gene Symbol: Nvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105980.1 | NP_001099450.1 | 19 | 2340 | ||
| XM_006250367.2 | XP_006250429.1 | 19 | 2264 | |||
| XM_006250368.2 | XP_006250430.1 | 19 | 2394 | |||
| XM_006250370.2 | XP_006250432.1 | 15 | 1973 | |||
| XM_006250372.2 | XP_006250434.1 | 15 | 1847 | |||

Nvl
Genomic Map