siRNA Details
| Gene Symbol: Ccdc121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223007.7 | XP_223007.4 | 1 | 509 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc121| Gene Symbol: Ccdc121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223007.7 | XP_223007.4 | 1 | 509 | ||