siRNA Details
| Gene Symbol: Ccdc185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170428.1 | NP_001163899.1 | 1 | 1306 | ||
| GenBank mRNA | BC098032.1 | - | 1 | 1306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc185| Gene Symbol: Ccdc185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170428.1 | NP_001163899.1 | 1 | 1306 | ||
| GenBank mRNA | BC098032.1 | - | 1 | 1306 | ||