siRNA Details
| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105982.1 | NP_001099452.1 | 8 | 1560 | ||
| XM_006250373.2 | XP_006250435.1 | 8 | 1720 | |||
| XM_006250375.2 | XP_006250437.1 | 8 | 1599 | |||
| XM_008769820.1 | XP_008768042.1 | 8 | 1720 | |||
| XM_008769821.1 | XP_008768043.1 | 8 | 1609 | |||
| XM_008769822.1 | XP_008768044.1 | 8 | 1698 | |||
| XM_008769823.1 | XP_008768045.1 | 8 | 1714 | |||
| GenBank mRNA | FQ212123.1 | - | 8 | 1544 | ||

Susd4
Genomic Map