siRNA Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105983.1 | NP_001099453.1 | 10 | 2393 | ||
| XM_006250381.2 | XP_006250443.1 | 9 | 2262 | |||
| XM_006250383.2 | XP_006250445.1 | 8 | 2723 | |||
| XM_006250384.2 | XP_006250446.1 | 8 | 2202 | |||
| XM_008769827.1 | XP_008768049.1 | 11 | 3087 | |||
| XM_008769828.1 | XP_008768050.1 | 9 | 2373 | |||

Disp1
Genomic Map