siRNA Details
| Gene Symbol: Lyplal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105986.1 | NP_001099456.1 | 2 | 218 | ||
| GenBank mRNA | BC166929.1 | - | 2 | 167 | ||
| FQ211186.1 | - | 2 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyplal1| Gene Symbol: Lyplal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105986.1 | NP_001099456.1 | 2 | 218 | ||
| GenBank mRNA | BC166929.1 | - | 2 | 167 | ||
| FQ211186.1 | - | 2 | 173 | |||