siRNA Details
Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105986.1 | NP_001099456.1 | 4 | 454 | ||
GenBank mRNA | FQ211186.1 | - | 4 | 409 | ||
FQ216980.1 | - | 1 | 37 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105986.1 | NP_001099456.1 | 4 | 454 | ||
GenBank mRNA | FQ211186.1 | - | 4 | 409 | ||
FQ216980.1 | - | 1 | 37 |