siRNA Details
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007617.1 | NP_001007618.1 | 1 | 177 | ||
GenBank mRNA | BC081899.1 | - | 1 | 177 | 7110518 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007617.1 | NP_001007618.1 | 1 | 177 | ||
GenBank mRNA | BC081899.1 | - | 1 | 177 | 7110518 |