siRNA Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105992.1 | NP_001099462.1 | 14 | 1937 | ||
| XM_008769626.1 | XP_008767848.1 | 14 | 2826 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpr1| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105992.1 | NP_001099462.1 | 14 | 1937 | ||
| XM_008769626.1 | XP_008767848.1 | 14 | 2826 | |||
Set Membership: |
|