siRNA Details
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037179.1 | NP_001032256.1 | 4 | 827 | ||
XM_006250542.2 | XP_006250604.1 | 2 | 560 | |||
GenBank mRNA | BC100076.1 | - | 4 | 827 | 7386007 |
