siRNA Details
| Gene Symbol: Lrrc8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037179.1 | NP_001032256.1 | 4 | 1206 | ||
| XM_006250542.2 | XP_006250604.1 | 2 | 939 | |||
| GenBank mRNA | BC100076.1 | - | 4 | 1206 | 7386007 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc8c| Gene Symbol: Lrrc8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037179.1 | NP_001032256.1 | 4 | 1206 | ||
| XM_006250542.2 | XP_006250604.1 | 2 | 939 | |||
| GenBank mRNA | BC100076.1 | - | 4 | 1206 | 7386007 | |