siRNA Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004209.1 | NP_001004209.1 | 3 | 388 | ||
| XM_006250627.2 | XP_006250689.1 | 4 | 575 | |||
| XM_006250629.2 | XP_006250691.1 | 4 | 499 | |||
| GenBank mRNA | BC078929.1 | - | 3 | 388 | 7107084 | |
| FQ210820.1 | - | 3 | 404 | |||
| FQ211398.1 | - | 3 | 411 | |||
| FQ222122.1 | - | 3 | 401 | |||

Hsd17b11
Genomic Map