siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105995.1 | NP_001099465.1 | 8 | 1710 | ||
| XM_008770004.1 | XP_008768226.1 | 10 | 2045 | |||
| XM_008770005.1 | XP_008768227.1 | 10 | 1949 | |||
| XM_008770006.1 | XP_008768228.1 | 10 | 1887 | |||
| XM_008770007.1 | XP_008768229.1 | 9 | 1714 | |||

Klhl8
Genomic Map