siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105995.1 | NP_001099465.1 | 2 | 459 | ||
| XM_008770004.1 | XP_008768226.1 | 4 | 794 | |||
| XM_008770005.1 | XP_008768227.1 | 4 | 698 | |||
| XM_008770006.1 | XP_008768228.1 | 4 | 636 | |||
| XM_008770007.1 | XP_008768229.1 | 3 | 463 | |||

Klhl8
Genomic Map