siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105995.1 | NP_001099465.1 | 9, 10 | 2012 | ||
| XM_008770004.1 | XP_008768226.1 | 11, 12 | 2347 | |||
| XM_008770005.1 | XP_008768227.1 | 11, 12 | 2251 | |||
| XM_008770006.1 | XP_008768228.1 | 11, 12 | 2189 | |||
| XM_008770007.1 | XP_008768229.1 | 10, 11 | 2016 | |||

Klhl8
Genomic Map