siRNA Details
| Gene Symbol: Ccni | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105998.1 | NP_001099468.1 | 8 | 1335 | ||
| XM_006250715.2 | XP_006250777.1 | 7 | 1435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccni| Gene Symbol: Ccni | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105998.1 | NP_001099468.1 | 8 | 1335 | ||
| XM_006250715.2 | XP_006250777.1 | 7 | 1435 | |||